Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275304.1 | NP_001262233.1 | 4 - 5 | 532 | 67 | |
NM_143697.5 | NP_651954.3 | 4 - 5 | 532 | 67 | ||
NM_168984.5 | NP_730773.4 | 4 - 5 | 543 | 67 | ||
GenBank mRNA | AY070910.1 | - | 4 - 5 | 530 | 67 | |
U49721.1 | - | 4 - 5 | 522 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |