Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140199.4 | NP_648456.2 | 2 | 1904 | 101 | |
NM_206321.2 | NP_996043.2 | 2 | 1904 | 101 | ||
GenBank mRNA | BT057991.1 | - | 2 | 1902 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |