Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nuf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038917.2 | NP_001034006.1 | 4 - 5 | 1630 | 55 | |
NM_001038918.1 | NP_001034007.1 | 5 - 6 | 904 | 55 | ||
NM_001038919.2 | NP_001034008.1 | 4 - 5 | 1764 | 55 | ||
NM_001038920.2 | NP_001034009.1 | 8 - 9 | 1977 | 55 | ||
NM_001038921.2 | NP_001034010.1 | 4 - 5 | 979 | 55 | ||
NM_001038922.2 | NP_001034011.1 | 8 - 9 | 1977 | 55 | ||
NM_001259819.2 | NP_001246748.1 | 6 - 7 | 1199 | 55 | ||
NM_001259820.2 | NP_001246749.1 | 9 - 10 | 2025 | 55 | ||
NM_001259821.2 | NP_001246750.1 | 9 - 10 | 2022 | 55 | ||
NM_001259822.2 | NP_001246751.1 | 4 - 5 | 1764 | 55 | ||
NM_001259823.2 | NP_001246752.1 | 4 - 5 | 1630 | 55 | ||
NM_001259824.1 | NP_001246753.1 | 8 - 9 | 1977 | 55 | ||
GenBank mRNA | AY069380.1 | - | 5 - 6 | 904 | 55 | |
BT011468.1 | - | 4 - 5 | 1628 | 55 | ||
BT100278.1 | - | 4 - 5 | 977 | 55 | ||
BT133122.1 | - | 8 - 9 | 1977 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |