Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169985.1 | NP_001163456.1 | 4 - 5 | 767 | 67 | |
NM_001202193.2 | NP_001189122.1 | 6 - 7 | 3454 | 67 | ||
NM_001259879.2 | NP_001246808.1 | 9 - 10 | 1858 | 67 | ||
NM_001275047.1 | NP_001261976.2 | 7 - 8 | 1643 | 67 | ||
NM_001316485.1 | NP_001303414.1 | 5 - 6 | 847 | 67 | ||
NM_140673.5 | NP_648930.1 | 9 - 10 | 1869 | 67 | ||
NM_140676.4 | NP_648933.3 | 4 - 5 | 763 | 67 | ||
NM_168694.3 | NP_730213.1 | 5 - 6 | 1052 | 67 | ||
NM_170635.3 | NP_730212.1 | 8 - 9 | 1813 | 67 | ||
NM_206387.3 | NP_996109.1 | 9 - 10 | 1757 | 67 | ||
NM_206388.3 | NP_996110.1 | 9 - 10 | 1981 | 67 | ||
NM_206389.3 | NP_996111.1 | 5 - 6 | 1063 | 67 | ||
NM_206390.2 | NP_996112.1 | 4 - 5 | 785 | 67 | ||
GenBank mRNA | AJ418713.1 | - | 9 - 10 | 1851 | 67 | |
AY069696.1 | - | 9 - 10 | 1851 | 67 | ||
AY071605.1 | - | 4 - 5 | 767 | 67 | ||
AY128470.1 | - | 4 - 5 | 785 | 67 | ||
BT009969.1 | - | 9 - 10 | 1866 | 67 | ||
BT044112.1 | - | 5 - 6 | 1051 | 67 | ||
BT100317.1 | - | 8 - 9 | 1811 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |