Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259888.3 | NP_001246817.1 | 3 - 4 | 673 | 61 | |
NM_080119.6 | NP_524858.1 | 4 - 5 | 839 | 61 | ||
NM_206396.4 | NP_996118.1 | 3 - 4 | 738 | 61 | ||
GenBank mRNA | AY118552.1 | - | 3 - 4 | 701 | 61 | |
X56689.1 | - | 824 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |