Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132092.3 | NP_572320.2 | 2 - 3 | 814 | 79 | |
NM_206634.2 | NP_996357.1 | 2 - 3 | 814 | 79 | ||
NM_206635.3 | NP_996358.2 | 2 - 3 | 653 | 79 | ||
GenBank mRNA | BT010027.1 | - | 2 - 3 | 814 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |