Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038746.3 | NP_001033835.2 | 8 - 9 | 3637 | 59 | |
NM_001038747.3 | NP_001033836.1 | 4 - 5 | 956 | 59 | ||
NM_001042798.2 | NP_001036263.1 | 2 - 3 | 491 | 59 | ||
NM_001258645.1 | NP_001245574.1 | 8 - 9 | 1818 | 59 | ||
NM_001258647.1 | NP_001245576.1 | 9 - 10 | 3157 | 59 | ||
NM_001272397.1 | NP_001259326.1 | 8 - 9 | 3637 | 59 | ||
NM_001272398.1 | NP_001259327.1 | 3 - 4 | 825 | 59 | ||
NM_001298081.1 | NP_001285010.1 | 7 - 8 | 2395 | 59 | ||
NM_001298082.1 | NP_001285011.1 | 7 - 8 | 2395 | 59 | ||
NM_132235.5 | NP_572463.1 | 4 - 5 | 956 | 59 | ||
NM_132236.6 | NP_572464.4 | 3 - 4 | 683 | 59 | ||
NM_206653.3 | NP_996376.1 | 3 - 4 | 618 | 59 | ||
GenBank mRNA | BT010281.1 | - | 4 - 5 | 954 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |