Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp10D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144715.2 | NP_001138187.1 | 3 - 4 | 984 | 85 | |
NM_001272524.1 | NP_001259453.1 | 3 - 4 | 812 | 85 | ||
NM_001272525.1 | NP_001259454.1 | 4 - 5 | 857 | 85 | ||
NM_167292.3 | NP_727544.2 | 3 - 4 | 812 | 85 | ||
NM_206690.2 | NP_996413.2 | 3 - 4 | 889 | 85 | ||
NM_206691.3 | NP_996414.2 | 3 - 4 | 1061 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |