Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014732.4 | NP_001014732.1 | 3 - 4 | 806 | 108 | |
NM_001014733.5 | NP_001014733.1 | 3 - 4 | 806 | 108 | ||
NM_001014734.4 | NP_001014734.1 | 3 - 4 | 806 | 108 | ||
NM_001014735.4 | NP_001014735.1 | 3 - 4 | 806 | 108 | ||
NM_001258707.2 | NP_001245636.1 | 3 - 4 | 806 | 108 | ||
NM_001258708.2 | NP_001245637.1 | 3 - 4 | 806 | 108 | ||
NM_001258709.2 | NP_001245638.1 | 2 - 3 | 733 | 108 | ||
NM_001258710.2 | NP_001245639.1 | 3 - 4 | 806 | 108 | ||
NM_001272548.1 | NP_001259477.1 | 2 - 3 | 736 | 108 | ||
NM_001370017.1 | NP_001356943.1 | 3 - 4 | 806 | 108 | ||
NM_001370018.1 | NP_001356944.1 | 3 - 4 | 806 | 108 | ||
NM_001370019.1 | NP_001356945.1 | 3 - 4 | 806 | 108 | ||
NM_078578.5 | NP_511133.2 | 3 - 4 | 806 | 108 | ||
NM_206693.5 | NP_996416.1 | 3 - 4 | 806 | 108 | ||
NM_206694.4 | NP_996417.1 | 3 - 4 | 816 | 108 | ||
NM_206695.4 | NP_996418.1 | 3 - 4 | 806 | 108 | ||
NM_206696.6 | NP_996419.1 | 3 - 4 | 806 | 108 | ||
NM_206697.4 | NP_996420.1 | 3 - 4 | 806 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |