Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078596.3 | NP_511151.2 | 2 - 3 | 948 | 75 | |
NM_206710.2 | NP_996433.1 | 2 - 3 | 948 | 75 | ||
GenBank mRNA | BT150470.1 | - | 2 - 3 | 943 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |