Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078613.4 | NP_511168.3 | 10 - 11 | 1905 | 83 | |
NM_206724.2 | NP_996447.1 | 10 - 11 | 2060 | 83 | ||
GenBank mRNA | AF132025.1 | - | 10 - 11 | 1909 | 83 | |
BT021401.1 | - | 10 - 11 | 1909 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |