Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042813.2 | NP_001036278.1 | 14 - 15 | 3031 | 71 | |
NM_001042814.2 | NP_001036279.1 | 14 - 15 | 3043 | 71 | ||
NM_001169297.2 | NP_001162768.1 | 14 - 15 | 2920 | 71 | ||
NM_001272660.2 | NP_001259589.1 | 15 - 16 | 2891 | 71 | ||
NM_080114.4 | NP_524853.2 | 13 - 14 | 2600 | 71 | ||
NM_167471.5 | NP_727911.1 | 14 - 15 | 2902 | 71 | ||
NM_206744.3 | NP_996467.1 | 14 - 15 | 2713 | 71 | ||
NM_206745.3 | NP_996468.1 | 14 - 15 | 2712 | 71 | ||
GenBank mRNA | X59435.1 | - | 2618 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |