Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206746.2 | NP_996469.1 | 6 - 7 | 1384 | 115 | |
GenBank mRNA | AY060660.1 | - | 5 - 6 | 2189 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |