Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144731.3 | NP_001138203.1 | 2 - 3 | 720 | 74 | |
NM_001298393.1 | NP_001285322.1 | 4 - 5 | 761 | 74 | ||
NM_080715.4 | NP_542446.2 | 4 - 5 | 1354 | 74 | ||
NM_167502.3 | NP_727959.2 | 3 - 4 | 1418 | 74 | ||
NM_167503.3 | NP_727960.1 | 4 - 5 | 833 | 74 | ||
NM_167504.2 | NP_727961.1 | 4 - 5 | 758 | 74 | ||
NM_206762.3 | NP_996485.1 | 4 - 5 | 902 | 74 | ||
GenBank mRNA | AY060841.1 | - | 4 - 5 | 745 | 74 | |
BT126193.1 | - | 4 - 5 | 760 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |