Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fur2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169302.2 | NP_001162773.1 | 14 - 15 | 4885 | 90 | |
NM_001169303.2 | NP_001162774.1 | 16 - 17 | 4762 | 90 | ||
NM_078644.4 | NP_523368.2 | 15 - 16 | 4834 | 90 | ||
NM_167506.3 | NP_727963.1 | 15 - 16 | 4843 | 90 | ||
NM_167507.3 | NP_727964.1 | 15 - 16 | 4943 | 90 | ||
NM_167508.2 | NP_727965.1 | 15 - 16 | 4877 | 90 | ||
NM_167509.3 | NP_727966.1 | 15 - 16 | 4937 | 90 | ||
NM_167510.3 | NP_727967.1 | 16 - 17 | 5054 | 90 | ||
NM_206763.2 | NP_996486.1 | 15 - 16 | 4821 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |