Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132947.5 | NP_573175.2 | 3 - 4 | 293 | 67 | |
NM_206770.2 | NP_996493.1 | 3 - 4 | 293 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |