Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201739.2 | NP_001188668.1 | 10 - 11 | 3565 | 76 | |
NM_001258798.2 | NP_001245727.1 | 12 - 13 | 3172 | 76 | ||
NM_001272736.2 | NP_001259665.2 | 9 - 10 | 1753 | 76 | ||
NM_001272737.1 | NP_001259666.2 | 9 - 10 | 2528 | 76 | ||
NM_078669.5 | NP_523393.3 | 9 - 10 | 1818 | 76 | ||
NM_167592.4 | NP_728120.1 | 9 - 10 | 1352 | 76 | ||
NM_167594.4 | NP_728122.1 | 10 - 11 | 3565 | 76 | ||
NM_167595.4 | NP_728123.1 | 9 - 10 | 1818 | 76 | ||
NM_206775.5 | NP_996498.2 | 12 - 13 | 3172 | 76 | ||
GenBank mRNA | BT050432.1 | - | 9 - 10 | 1352 | 76 | |
X07132.1 | - | 1446 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |