Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ari-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258807.3 | NP_001245736.1 | 2 - 3 | 714 | 92 | |
NM_001272742.2 | NP_001259671.1 | 2 - 3 | 809 | 92 | ||
NM_001272743.1 | NP_001259672.1 | 2 - 3 | 925 | 92 | ||
NM_078675.4 | NP_523399.1 | 2 - 3 | 714 | 92 | ||
NM_167610.4 | NP_728145.1 | 1 - 2 | 693 | 92 | ||
NM_206777.2 | NP_996500.1 | 2 - 3 | 869 | 92 | ||
GenBank mRNA | X98310.1 | - | 663 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |