Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ari-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258807.3 | NP_001245736.1 | 5 - 6 | 1581 | 73 | |
NM_001272742.2 | NP_001259671.1 | 5 - 6 | 1676 | 73 | ||
NM_001272743.1 | NP_001259672.1 | 5 - 6 | 1792 | 73 | ||
NM_078675.4 | NP_523399.1 | 5 - 6 | 1581 | 73 | ||
NM_167610.4 | NP_728145.1 | 4 - 5 | 1560 | 73 | ||
NM_206777.2 | NP_996500.1 | 5 - 6 | 1736 | 73 | ||
GenBank mRNA | X98310.1 | - | 1530 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |