Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbp21E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134700.4 | NP_608544.1 | 1 - 2 | 916 | 86 | |
GenBank mRNA | AY058512.1 | - | 1 - 2 | 868 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |