Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chinmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201751.2 | NP_001188680.1 | 3 - 4 | 2153 | 69 | |
NM_001258910.2 | NP_001245839.1 | 3 - 4 | 2153 | 69 | ||
NM_134766.6 | NP_608610.1 | 2 - 3 | 772 | 69 | ||
NM_164428.3 | NP_722717.2 | 4 - 5 | 870 | 69 | ||
NM_164429.3 | NP_722718.1 | 4 - 5 | 1922 | 69 | ||
NM_164430.4 | NP_722719.1 | 3 - 4 | 1852 | 69 | ||
GenBank mRNA | AY060464.1 | - | 4 - 5 | 879 | 69 | |
BT050405.1 | - | 3 - 4 | 1851 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |