Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272978.1 | NP_001259907.1 | 4 - 5 | 1006 | 73 | |
NM_134805.4 | NP_608649.2 | 4 - 5 | 814 | 73 | ||
NM_164456.3 | NP_722765.2 | 4 - 5 | 1009 | 73 | ||
GenBank mRNA | BT044192.1 | - | 4 - 5 | 778 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |