Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134812.3 | NP_608656.1 | 1 - 2 | 257 | 64 | |
GenBank mRNA | AY061485.1 | - | 1 - 2 | 184 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |