Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hydr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258923.2 | NP_001245852.1 | 3 - 4 | 959 | 86 | |
NM_001273064.1 | NP_001259993.1 | 4 - 5 | 1083 | 86 | ||
NM_134907.4 | NP_608751.2 | 3 - 4 | 959 | 86 | ||
GenBank mRNA | BT010082.1 | - | 3 - 4 | 946 | 86 | |
BT124766.1 | - | 4 - 5 | 1063 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |