Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134983.3 | NP_608827.1 | 2 - 3 | 1067 | 83 | |
GenBank mRNA | AF145653.2 | - | 2 - 3 | 1029 | 83 | |
BT021328.1 | - | 2 - 3 | 1073 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |