Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140023.3 | NP_648280.1 | 4 - 5 | 787 | 68 | |
GenBank mRNA | AY071335.1 | - | 4 - 5 | 785 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |