Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 2 - 3 | 743 | 68 | |
NM_140031.3 | NP_648288.2 | 2 - 3 | 743 | 68 | ||
NM_168315.3 | NP_729452.1 | 2 - 3 | 743 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |