Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nwk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259760.2 | NP_001246689.1 | 5 - 6 | 1787 | 68 | |
NM_001259761.2 | NP_001246690.1 | 5 - 6 | 1787 | 68 | ||
NM_001259762.2 | NP_001246691.1 | 5 - 6 | 1862 | 68 | ||
NM_001259763.2 | NP_001246692.1 | 5 - 6 | 1787 | 68 | ||
NM_001300064.1 | NP_001286993.1 | 5 - 6 | 1787 | 68 | ||
NM_001300065.1 | NP_001286994.1 | 5 - 6 | 1787 | 68 | ||
NM_001300066.1 | NP_001286995.1 | 5 - 6 | 1787 | 68 | ||
NM_001300067.1 | NP_001286996.1 | 5 - 6 | 1862 | 68 | ||
NM_001300068.1 | NP_001286997.1 | 5 - 6 | 1787 | 68 | ||
NM_140033.3 | NP_648290.3 | 5 - 6 | 1787 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |