Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iPLA2-VIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140109.2 | NP_648366.2 | 6 - 7 | 2549 | 85 | |
NM_168383.2 | NP_729565.2 | 7 - 8 | 2687 | 85 | ||
NM_168384.2 | NP_729566.2 | 6 - 7 | 2965 | 85 | ||
NM_168385.2 | NP_729567.2 | 7 - 8 | 2872 | 85 | ||
GenBank mRNA | BT125951.1 | - | 6 - 7 | 2426 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |