Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: simj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274755.1 | NP_001261684.1 | 5 - 6 | 2025 | 55 | |
NM_001274756.1 | NP_001261685.1 | 5 - 6 | 2001 | 55 | ||
NM_140150.3 | NP_648407.1 | 5 - 6 | 2001 | 55 | ||
NM_168420.2 | NP_729624.1 | 5 - 6 | 2001 | 55 | ||
NM_168421.2 | NP_729625.1 | 6 - 7 | 1760 | 55 | ||
GenBank mRNA | AY051828.1 | - | 5 - 6 | 1966 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |