Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JIL-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274768.1 | NP_001261697.1 | 2 - 3 | 1796 | 88 | |
NM_140175.3 | NP_648432.1 | 2 - 3 | 1796 | 88 | ||
NM_168439.2 | NP_729661.1 | 1 - 2 | 958 | 88 | ||
GenBank mRNA | AF142061.1 | - | 2 - 3 | 1751 | 88 | |
BT044484.1 | - | 1 - 2 | 950 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |