Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JIL-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274768.1 | NP_001261697.1 | 6 - 7 | 2896 | 79 | |
NM_140175.3 | NP_648432.1 | 6 - 7 | 2896 | 79 | ||
NM_168439.2 | NP_729661.1 | 5 - 6 | 2058 | 79 | ||
GenBank mRNA | AF142061.1 | - | 6 - 7 | 2851 | 79 | |
BT044484.1 | - | 5 - 6 | 2050 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |