Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP68F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274814.1 | NP_001261743.1 | 4 - 5 | 413 | 66 | |
NM_001274815.1 | NP_001261744.1 | 3 - 4 | 447 | 66 | ||
NM_140295.3 | NP_648552.1 | 3 - 4 | 381 | 66 | ||
GenBank mRNA | AY069311.1 | - | 3 - 4 | 368 | 66 | |
BT126081.1 | - | 4 - 5 | 389 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |