Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tral | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274840.1 | NP_001261769.1 | 5 - 6 | 2006 | 81 | |
NM_001274841.1 | NP_001261770.1 | 5 - 6 | 2018 | 81 | ||
NM_001274842.1 | NP_001261771.1 | 5 - 6 | 1988 | 81 | ||
NM_001274843.1 | NP_001261772.1 | 5 - 6 | 1976 | 81 | ||
NM_140328.5 | NP_648585.3 | 5 - 6 | 2006 | 81 | ||
GenBank mRNA | AY069066.1 | - | 5 - 6 | 1988 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |