Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169962.2 | NP_001163433.1 | 8 - 9 | 2891 | 62 | |
NM_140344.3 | NP_648601.1 | 9 - 10 | 2831 | 62 | ||
GenBank mRNA | AY129445.1 | - | 8 - 9 | 2860 | 62 | |
BT021340.1 | - | 9 - 10 | 2766 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |