Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICAL-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140364.3 | NP_648621.1 | 1 | 3265 | 98 | |
GenBank mRNA | AF520716.1 | - | 1 | 3240 | 98 | |
AY051991.1 | - | 1 | 3240 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |