Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057864.4 | NP_477212.1 | 3 - 4 | 3453 | 73 | |
NM_164675.2 | NP_723146.1 | 3 - 4 | 3509 | 73 | ||
NM_164676.2 | NP_723147.1 | 2 - 3 | 3449 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |