Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258983.2 | NP_001245912.1 | 3 - 4 | 900 | 73 | |
NM_001273193.1 | NP_001260122.1 | 6 - 7 | 1177 | 73 | ||
NM_164682.3 | NP_723156.1 | 4 - 5 | 641 | 73 | ||
NM_164683.3 | NP_723157.1 | 4 - 5 | 1005 | 73 | ||
NM_164684.3 | NP_723158.1 | 4 - 5 | 1005 | 73 | ||
GenBank mRNA | AF202774.1 | - | 4 - 5 | 563 | 73 | |
AY061071.1 | - | 4 - 5 | 991 | 73 | ||
BT001751.1 | - | 4 - 5 | 640 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |