Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103635.3 | NP_001097105.3 | 5 - 6 | 1386 | 72 | |
NM_001144329.2 | NP_001137801.1 | 5 - 6 | 1379 | 72 | ||
NM_001273217.1 | NP_001260146.1 | 5 - 6 | 879 | 72 | ||
NM_001273218.1 | NP_001260147.1 | 5 - 6 | 1379 | 72 | ||
NM_135220.3 | NP_609064.3 | 5 - 6 | 1379 | 72 | ||
NM_164701.2 | NP_723202.2 | 5 - 6 | 1379 | 72 | ||
GenBank mRNA | BT124889.1 | - | 5 - 6 | 879 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |