Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: homer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042876.2 | NP_001036341.2 | 3 - 4 | 894 | 70 | |
NM_001273221.1 | NP_001260150.1 | 3 - 4 | 894 | 70 | ||
NM_001273222.1 | NP_001260151.1 | 3 - 4 | 870 | 70 | ||
NM_001273223.1 | NP_001260152.1 | 3 - 4 | 903 | 70 | ||
NM_058048.4 | NP_477396.1 | 3 - 4 | 894 | 70 | ||
NM_164706.2 | NP_723207.1 | 3 - 4 | 861 | 70 | ||
GenBank mRNA | AF093266.1 | - | 3 - 4 | 882 | 70 | |
BT004910.1 | - | 3 - 4 | 873 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |