Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273248.1 | NP_001260177.1 | 4 - 5 | 1289 | 72 | |
NM_001273249.1 | NP_001260178.1 | 4 - 5 | 1289 | 72 | ||
NM_078777.3 | NP_523501.1 | 3 - 4 | 889 | 72 | ||
NM_164743.3 | NP_723263.2 | 4 - 5 | 1096 | 72 | ||
NM_164744.2 | NP_723264.1 | 4 - 5 | 1096 | 72 | ||
GenBank mRNA | AF047468.1 | - | 3 - 4 | 631 | 72 | |
BT128745.1 | - | 4 - 5 | 1093 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |