Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273260.1 | NP_001260189.1 | 3 - 4 | 1299 | 100 | |
NM_001273261.1 | NP_001260190.1 | 3 - 4 | 1299 | 100 | ||
NM_078779.3 | NP_523503.2 | 3 - 4 | 1333 | 100 | ||
NM_164747.1 | NP_723271.1 | 3 - 4 | 1299 | 100 | ||
GenBank mRNA | BT150399.1 | - | 3 - 4 | 1317 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |