Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038792.1 | NP_001033881.1 | 4 - 5 | 1131 | 80 | |
NM_001201781.1 | NP_001188710.1 | 5 - 6 | 1182 | 80 | ||
NM_001273282.1 | NP_001260211.1 | 4 - 5 | 1131 | 80 | ||
NM_058001.3 | NP_477349.1 | 4 - 5 | 1131 | 80 | ||
NM_164756.3 | NP_723292.1 | 6 - 7 | 1558 | 80 | ||
NM_164757.2 | NP_723293.1 | 5 - 6 | 1195 | 80 | ||
GenBank mRNA | BT021228.1 | - | 4 - 5 | 1132 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |