Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038792.1 | NP_001033881.1 | 5 - 6 | 1336 | 72 | |
NM_001201781.1 | NP_001188710.1 | 6 - 7 | 1387 | 72 | ||
NM_001201782.1 | NP_001188711.1 | 7 - 8 | 1440 | 72 | ||
NM_001201783.1 | NP_001188712.1 | 4 - 5 | 1168 | 72 | ||
NM_001201784.2 | NP_001188713.1 | 5 - 6 | 1232 | 72 | ||
NM_001273282.1 | NP_001260211.1 | 5 - 6 | 1336 | 72 | ||
NM_001273283.1 | NP_001260212.1 | 6 - 7 | 1595 | 72 | ||
NM_058001.3 | NP_477349.1 | 5 - 6 | 1336 | 72 | ||
NM_135287.2 | NP_609131.2 | 4 - 5 | 1011 | 72 | ||
NM_164756.3 | NP_723292.1 | 7 - 8 | 1763 | 72 | ||
NM_164757.2 | NP_723293.1 | 6 - 7 | 1400 | 72 | ||
GenBank mRNA | BT021228.1 | - | 5 - 6 | 1337 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |