Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078782.3 | NP_523506.1 | 6 - 7 | 2451 | 64 | |
NM_164761.2 | NP_723298.1 | 6 - 7 | 2388 | 64 | ||
NM_164762.2 | NP_723299.2 | 6 - 7 | 2418 | 64 | ||
NM_164763.2 | NP_723300.2 | 6 - 7 | 2391 | 64 | ||
NM_164764.2 | NP_723301.1 | 6 - 7 | 2415 | 64 | ||
GenBank mRNA | AJ269539.1 | - | 6 - 7 | 2435 | 64 | |
AY122084.1 | - | 6 - 7 | 2435 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |