Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273295.1 | NP_001260224.1 | 6 - 7 | 1296 | 62 | |
NM_164782.2 | NP_723338.1 | 6 - 7 | 1154 | 62 | ||
NM_164783.2 | NP_723339.1 | 6 - 7 | 1039 | 62 | ||
NM_164784.2 | NP_723340.1 | 6 - 7 | 1363 | 62 | ||
NM_164785.2 | NP_723341.1 | 6 - 7 | 1248 | 62 | ||
NM_164786.2 | NP_723342.1 | 6 - 7 | 1087 | 62 | ||
NM_164787.2 | NP_723343.1 | 7 - 8 | 1392 | 62 | ||
NM_164788.2 | NP_723344.1 | 6 - 7 | 1296 | 62 | ||
NM_164789.3 | NP_723345.1 | 5 - 6 | 1228 | 62 | ||
NM_164790.3 | NP_723346.2 | 8 - 9 | 2275 | 62 | ||
GenBank mRNA | BT005198.1 | - | 6 - 7 | 1001 | 62 | |
BT006333.1 | - | 6 - 7 | 1203 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |