Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164790.3 | NP_723346.2 | 9 - 10 | 2393 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |