Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piezo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042881.5 | NP_001036346.3 | 19 - 20 | 5755 | 55 | |
NM_001169436.3 | NP_001162907.1 | 18 - 19 | 5533 | 55 | ||
NM_001201789.2 | NP_001188718.1 | 18 - 19 | 5338 | 55 | ||
NM_001201790.2 | NP_001188719.1 | 17 - 18 | 5281 | 55 | ||
NM_001273297.1 | NP_001260226.1 | 19 - 20 | 5608 | 55 | ||
NM_001273298.1 | NP_001260227.1 | 17 - 18 | 5281 | 55 | ||
NM_001273299.1 | NP_001260228.1 | 18 - 19 | 5530 | 55 | ||
NM_001273300.2 | NP_001260229.2 | 18 - 19 | 6142 | 55 | ||
NM_001316385.1 | NP_001303314.1 | 19 - 20 | 6394 | 55 | ||
NM_135344.6 | NP_609188.4 | 18 - 19 | 5277 | 55 | ||
NM_164795.6 | NP_723354.3 | 19 - 20 | 5755 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |