Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169438.2 | NP_001162909.1 | 14 - 15 | 4328 | 79 | |
NM_001169439.2 | NP_001162910.1 | 15 - 16 | 4020 | 79 | ||
NM_001169440.2 | NP_001162911.1 | 15 - 16 | 4020 | 79 | ||
NM_001201791.2 | NP_001188720.1 | 13 - 14 | 3789 | 79 | ||
NM_001201792.2 | NP_001188721.1 | 14 - 15 | 3689 | 79 | ||
NM_001273305.1 | NP_001260234.1 | 14 - 15 | 3993 | 79 | ||
NM_001273306.1 | NP_001260235.1 | 13 - 14 | 3786 | 79 | ||
NM_078785.4 | NP_523509.2 | 14 - 15 | 3816 | 79 | ||
NM_164801.3 | NP_723365.1 | 14 - 15 | 3816 | 79 | ||
NM_175983.3 | NP_787997.2 | 14 - 15 | 3693 | 79 | ||
NM_205925.3 | NP_995647.2 | 13 - 14 | 3786 | 79 | ||
NM_205926.3 | NP_995648.1 | 14 - 15 | 3851 | 79 | ||
NM_205927.3 | NP_995649.1 | 14 - 15 | 4433 | 79 | ||
GenBank mRNA | BT050546.1 | - | 13 - 14 | 3460 | 79 | |
BT133154.1 | - | 15 - 16 | 3989 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |