Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mur29B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164808.4 | NP_723377.1 | 1 - 2 | 184 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |